Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unkl
Entrez Gene ID
100144682
Gene Name
unkempt family like zinc finger
Gene Aliases
zc3h5l, zc3hdc5l
Chromosome Location
Chr.9: 22420134 - 22472912 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123443.1 NP_001116915.1 2 - 3 428 92
XM_012970032.2 XP_012825486.1 2 - 3 1849 92
GenBank mRNA BC160454.1 - 2 - 3 428 92

View Full Product Details