Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kremen1
Entrez Gene ID
100144698
Gene Name
kringle containing transmembrane protein 1
Gene Aliases
kremem1, kremen, krm1
Chromosome Location
Chr.1: 157478892 - 157561124 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123455.1 NP_001116927.1 5 - 6 815 104
XM_012955367.2 XP_012810821.1 5 - 6 962 104
GenBank mRNA BC161114.1 - 5 - 6 815 104

View Full Product Details