Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gal3st4
Entrez Gene ID
100145173
Gene Name
galactose-3-O-sulfotransferase 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683390: 314693 - 332010 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126666.1 NP_001120138.1 3 - 4 592 79
XM_012953352.2 XP_012808806.1 3 - 4 821 79
XM_012953354.1 XP_012808808.1 3 - 4 506 79
XM_012953355.2 XP_012808809.1 3 - 4 1277 79
XM_012953356.2 XP_012808810.1 3 - 4 413 79
GenBank mRNA BC159090.1 - 3 - 4 592 79
CU075328.1 - 3 - 4 500 79

View Full Product Details