Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
il21
Entrez Gene ID
100145684
Gene Name
interleukin 21
Gene Aliases
il-21
Chromosome Location
Chr.1: 56598002 - 56605963 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127059.1 NP_001120531.1 2 - 3 172 75
GenBank mRNA EF533945.1 - 2 - 3 172 75

View Full Product Details