Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
abhd6
Entrez Gene ID
100145783
Gene Name
abhydrolase domain containing 6
Gene Aliases
abhd6-a, abhd6-b
Chromosome Location
Chr.Un|NW_016683423: 401902 - 411274 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127147.1 NP_001120619.1 2 - 3 368 67
GenBank mRNA BC161759.1 - 2 - 3 368 67

View Full Product Details