Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnajc8
Entrez Gene ID
100145281
Gene Name
DnaJ heat shock protein family (Hsp40) member C8
Gene Aliases
-
Chromosome Location
Chr.2: 53467765 - 53486728 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126759.1 NP_001120231.1 4 - 5 335 69
GenBank mRNA BC160408.1 - 4 - 5 335 69

View Full Product Details