Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dse
Entrez Gene ID
100145321
Gene Name
dermatan sulfate epimerase
Gene Aliases
DS-epi1, SART2
Chromosome Location
Chr.5: 71785570 - 71806749 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126796.1 NP_001120268.1 3 - 4 877 85
XM_012962729.2 XP_012818183.1 2 - 3 338 85
GenBank mRNA BC160517.1 - 3 - 4 877 85
CR942332.2 - 3 - 4 769 85

Gene Symbol
LOC101731195
Entrez Gene ID
101731195
Gene Name
dermatan-sulfate epimerase-like
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683805: 27121 - 33314 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004919990.2 XP_004920047.2 1 - 2 1138 85

View Full Product Details