Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nuak2
Entrez Gene ID
100158605
Gene Name
NUAK family, SNF1-like kinase, 2
Gene Aliases
-
Chromosome Location
Chr.2: 64631468 - 64645742 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128027.1 NP_001121499.1 3 - 4 465 58
GenBank mRNA BC166319.1 - 3 - 4 465 58

View Full Product Details