Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wnt11
Entrez Gene ID
100158655
Gene Name
Wnt family member 11
Gene Aliases
Wnt11-R, Xwnt11, wnt11r
Chromosome Location
Chr.2: 169123862 - 169171926 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128058.1 NP_001121530.1 2 - 3 478 80
GenBank mRNA BC166399.1 - 2 - 3 478 80

View Full Product Details