The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129928.1 | NP_001123400.1 | 6 - 7 | 1202 | 76 |
GenBank mRNA | BC167362.1 | - | 6 - 7 | 1202 | 76 |