Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
btbd1
Entrez Gene ID
100170424
Gene Name
BTB (POZ) domain containing 1
Gene Aliases
ns5atp8
Chromosome Location
Chr.3: 95461804 - 95506435 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130202.1 NP_001123674.1 2 - 3 336 81
XM_018091949.1 XP_017947438.1 3 - 4 656 81
GenBank mRNA BC166965.1 - 2 - 3 336 81

View Full Product Details