Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bcr
Entrez Gene ID
100170543
Gene Name
BCR, RhoGEF and GTPase activating protein
Gene Aliases
bcr1, cml, phl
Chromosome Location
Chr.1: 143920826 - 143951468 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130320.1 NP_001123792.1 10 - 11 2716 87
GenBank mRNA BC167580.1 - 10 - 11 2716 87

View Full Product Details