Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
brd2
Entrez Gene ID
100038037
Gene Name
bromodomain containing 2
Gene Aliases
d6s113e, fsh, fsrg1, nat, ring3, rnf3
Chromosome Location
Chr.8: 52010973 - 52026265 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134810.1 NP_001128282.1 4 - 5 702 102
XM_012953823.2 XP_012809277.1 4 - 5 1232 102
XM_012953824.2 XP_012809278.1 4 - 5 1415 102
XM_012953825.2 XP_012809279.1 5 - 6 1334 102
XM_018096085.1 XP_017951574.1 4 - 5 872 102
GenBank mRNA BC168574.1 - 4 - 5 702 102
CR762100.2 - 4 - 5 678 102

View Full Product Details