Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
brd2
Entrez Gene ID
100038037
Gene Name
bromodomain containing 2
Gene Aliases
d6s113e, fsh, fsrg1, nat, ring3, rnf3
Chromosome Location
Chr.8: 52010973 - 52026265 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134810.1 NP_001128282.1 5 - 6 915 84
XM_012953823.2 XP_012809277.1 5 - 6 1445 84
XM_012953824.2 XP_012809278.1 5 - 6 1628 84
XM_012953825.2 XP_012809279.1 6 - 7 1547 84
XM_018096085.1 XP_017951574.1 5 - 6 1085 84
GenBank mRNA BC168574.1 - 5 - 6 915 84
CR762100.2 - 5 - 6 891 84

View Full Product Details