Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slc9a3r2
Entrez Gene ID
100216077
Gene Name
SLC9A3 regulator 2
Gene Aliases
e3karp, nhe3rf2, nherf-2, nherf2, octs2, sip-1, sip1, tka-1
Chromosome Location
Chr.9: 12960973 - 13031812 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142062.1 NP_001135534.1 4 - 5 1071 74
GenBank mRNA BC168002.1 - 4 - 5 1071 74

View Full Product Details