Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sirt1
Entrez Gene ID
100145679
Gene Name
sirtuin 1
Gene Aliases
sir2l1, xsirt1
Chromosome Location
Chr.7: 30060155 - 30074245 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142909.1 NP_001136381.1 2 - 3 615 110
XM_012966096.2 XP_012821550.1 3 - 4 1135 110
GenBank mRNA BC161420.1 - 2 - 3 615 110
CR926386.2 - 3 - 4 699 110

View Full Product Details