Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tmcc1
Entrez Gene ID
100170168
Gene Name
transmembrane and coiled-coil domain family 1
Gene Aliases
-
Chromosome Location
Chr.4: 121603287 - 121687392 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142914.1 NP_001136386.1 6 - 7 4207 104
XM_012960654.2 XP_012816108.1 7 - 8 2089 104
XM_012960655.2 XP_012816109.1 6 - 7 1997 104
XM_012960656.1 XP_012816110.1 4 - 5 1236 104
XM_012960657.2 XP_012816111.1 4 - 5 1198 104
XM_012960658.2 XP_012816112.1 4 - 5 1324 104
XM_012960659.1 XP_012816113.1 4 - 5 1346 104
XM_012960660.2 XP_012816114.1 4 - 5 1418 104
XM_012960662.2 XP_012816116.1 4 - 5 1261 104
XM_018092939.1 XP_017948428.1 8 - 9 2242 104
GenBank mRNA BC167584.1 - 6 - 7 4207 104
CT025256.2 - 6 - 7 1984 104

View Full Product Details