Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tinf2
Entrez Gene ID
100233216
Gene Name
TERF1 (TRF1)-interacting nuclear factor 2
Gene Aliases
tin2
Chromosome Location
Chr.4: 61958796 - 61965125 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143925.1 NP_001137397.1 3 - 4 326 93
XM_018092949.1 XP_017948438.1 3 - 4 509 93
GenBank mRNA CR761197.2 - 3 - 4 421 93
EU422982.1 - 3 - 4 326 93

View Full Product Details