The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005097.2 | NP_001005097.1 | 5 - 5 | 753 | 75 |
GenBank mRNA | BC077029.1 | - | 5 - 5 | 754 | 75 |
CR761734.2 | - | 5 - 5 | 732 | 75 | |
CR761793.2 | - | 5 - 5 | 730 | 75 |