Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
armc7
Entrez Gene ID
100216038
Gene Name
armadillo repeat containing 7
Gene Aliases
-
Chromosome Location
Chr.10: 35774587 - 35778813 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142026.1 NP_001135498.1 2 - 2 552 112
XM_012971561.2 XP_012827015.1 3 - 3 604 112
XM_018097677.1 XP_017953166.1 3 - 3 659 112
GenBank mRNA BC167900.1 - 2 - 2 552 112

View Full Product Details