Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
sid1
Entrez Gene ID
100271892
Gene Name
sister of indeterminate spikelet 1
Gene Aliases
ZEAMMB73_275310
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146067.1 NP_001139539.1 - 1137 85
GenBank mRNA BT063357.1 - - 1137 85

View Full Product Details