Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
RH2
Entrez Gene ID
732749
Gene Name
RH2 protein-like
Gene Aliases
ZEAMMB73_174730
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112378.1 NP_001105848.1 - 1185 107
GenBank mRNA BT039346.1 - - 1097 107
DQ327709.1 - - 1185 107

View Full Product Details