Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
rnc1
Entrez Gene ID
100125654
Gene Name
ribonuclease III domain protein1
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112585.1 NP_001106055.1 - 863 117
GenBank mRNA BT041100.1 - - 776 117
BT064625.1 - - 842 117
EF650835.1 - - 863 117
EU971053.1 - - 805 117

View Full Product Details