Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LOC100280873
Entrez Gene ID
100280873
Gene Name
protein transport protein Sec61 beta subunit
Gene Aliases
ZEAMMB73_322940
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001153793.1 NP_001147265.1 - 748 76
GenBank mRNA EU954339.1 - - 748 76

View Full Product Details