Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
rgd1
Entrez Gene ID
100037819
Gene Name
ragged seedling1
Gene Aliases
SGS3, ZEAMMB73_663370, lbl1
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112519.1 NP_001105989.1 - 137 84
GenBank mRNA BT066397.1 - - 84
DQ832257.1 - - 137 84

View Full Product Details