Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
cesa5
Entrez Gene ID
541803
Gene Name
cellulose synthase5
Gene Aliases
CesA-5
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111485.1 NP_001104955.1 - 816 70
GenBank mRNA AF200529.1 - - 816 70
AY110079.1 - - 816 70

View Full Product Details