Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
elm1
Entrez Gene ID
542164
Gene Name
elongated mesocotyl1
Gene Aliases
Hy2
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111786.1 NP_001105256.1 - 752 73
GenBank mRNA AY560384.1 - - 752 73
EU956180.1 - - 732 73

View Full Product Details