Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ERD6
Entrez Gene ID
100193700
Gene Name
sugar transporter protein ERD6
Gene Aliases
ZEAMMB73_635976
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001138792.2 NP_001132264.2 - 1079 94
NM_001204251.1 NP_001191180.1 - 1079 94
GenBank mRNA BT036036.1 - - 410 94
BT064122.1 - - 1225 94
FJ750248.1 - - 1079 94
FJ750249.1 - - 1079 94

View Full Product Details