Gene Transcripts
- Gene Symbol
- Dcm5
- Entrez Gene ID
- 257653
- Gene Name
- Dcm5 protein
- Gene Aliases
- Chromosome Location
- Chr. 2: 233909142 - 233909909 on Build Rnor_6.0
- UniGene ID
- Rn.81019
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033150.1 |
|
1 |
525 |
|
GenBank mRNA |
AF412817.1 |
- |
1 |
526 |
|
View Full Product Details