Gene Transcripts
- Gene Symbol
- Mirg
- Entrez Gene ID
- 100040724
- Gene Name
- miRNA containing gene
- Gene Aliases
- 2810474H01Rik, AI852992, Gm2928, Meg9, mir-410
- Chromosome Location
- Chr. 12: 109734981 - 109749458 on Build GRCm38
- UniGene ID
- Mm.260543
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB193946.1 |
- |
1, 2 |
42 |
|
Chr. 12 - 109734985 - 109743827 [+] |
GenBankmRNA |
CB193969.1 |
- |
1, 2 |
42 |
|
Chr. 12 - 109734985 - 109748183 [+] |
GenBankmRNA |
BG805738.1 |
- |
1, 2 |
31 |
|
Chr. 12 - 109734996 - 109742244 [+] |
View Full Product Details