Gene Transcripts

Gene Symbol
Speer4cos
Entrez Gene ID
75858
Gene Name
spermatogenesis associated glutamate (E)-rich protein 4c, opposite strand transcript
Gene Aliases
4930568L21Rik, SPEER-7, Speer7-ps, Speer7-ps1
Chromosome Location
Chr. 5: 15680710 - 15714596 on Build GRCm38
UniGene ID
Mm.359463
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001585.3 5 762


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK016255.1 - 761 Chr. - - [-]

View Full Product Details