Gene Transcripts

Gene Symbol
Snhg8
Entrez Gene ID
69895
Gene Name
small nucleolar RNA host gene 8
Gene Aliases
1110065M07Rik, 2010109N14Rik
Chromosome Location
Chr. 3: 123507552 - 123508336 on Build GRCm38
UniGene ID
Mm.260791
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028574.1 1 612
GenBank mRNA AK008369.1 - 1 63
BC025067.1 - 2 359


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BU514542.1 - 596 Chr. - - [-]
GenBankmRNA CB181105.1 - 1 629 Chr. 3 - 123507562 - 123508330 [-]
GenBankmRNA CN693661.1 - 1 599 Chr. 3 - 123507686 - 123508323 [-]
GenBankmRNA BF181273.1 - 2 358 Chr. 3 - 123507550 - 123508320 [-]

View Full Product Details