Gene Transcripts
- Gene Symbol
- Snhg8
- Entrez Gene ID
- 69895
- Gene Name
- small nucleolar RNA host gene 8
- Gene Aliases
- 1110065M07Rik, 2010109N14Rik
- Chromosome Location
- Chr. 3: 123507552 - 123508336 on Build GRCm38
- UniGene ID
- Mm.260791
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB181105.1 |
- |
1 |
702 |
|
Chr. 3 - 123507562 - 123508330 [-] |
GenBankmRNA |
BF181273.1 |
- |
2 |
430 |
|
Chr. 3 - 123507550 - 123508320 [-] |
View Full Product Details