Gene Transcripts
- Gene Symbol
- Snord22
- Entrez Gene ID
- 100127111
- Gene Name
- small nucleolar RNA, C/D box 22
- Gene Aliases
- Rnu22, U22
- Chromosome Location
- Chr. 19: 8725866 - 8725991 on Build GRCm38
- UniGene ID
- Mm.292489
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_004445.1 |
|
1 |
19 |
|
- Gene Symbol
- Snhg1
- Entrez Gene ID
- 83673
- Gene Name
- small nucleolar RNA host gene 1
- Gene Aliases
- Chromosome Location
- Chr. 19: 8723487 - 8726326 on Build GRCm38
- UniGene ID
- Mm.292489
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK036616.1 |
- |
2 |
2171 |
|
AK051045.1 |
- |
3 |
1927 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AA590725.1 |
- |
2 |
143 |
|
Chr. 19 - 8725484 - 8726270 [+] |
GenBankmRNA |
AV576327.1 |
- |
2 |
127 |
|
Chr. 19 - 8725500 - 8726440 [+] |
GenBankmRNA |
AV502115.1 |
- |
3 |
376 |
|
Chr. 19 - 8725029 - 8726002 [+] |
GenBankmRNA |
CO813536.1 |
- |
2 |
668 |
|
Chr. 19 - 8725004 - 8725964 [+] |
GenBankmRNA |
U40654.1 |
- |
1 |
2555 |
|
Chr. - - [-] |
GenBankmRNA |
AA110623.1 |
- |
7 |
361 |
|
Chr. 19 - 8724170 - 8725927 [+] |
View Full Product Details