Gene Transcripts
- Gene Symbol
- Marcksl1-ps4
- Entrez Gene ID
- 111002
- Gene Name
- MARCKS-like 1, pseudogene 4
- Gene Aliases
- Gm16264, Mlp-rs4, Mrp-rs4, OTTMUSG00000032901
- Chromosome Location
- Chr. 13: 4248735 - 4249777 on Build GRCm38
- UniGene ID
- Mm.482672
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028405.1 |
|
1 |
807 |
|
GenBank mRNA |
BB624916.1 |
- |
1 |
279 |
|
View Full Product Details