Gene Transcripts
- Gene Symbol
- Vmn2r29
- Entrez Gene ID
- 76229
- Gene Name
- vomeronasal 2, receptor 29
- Gene Aliases
- 6430701C03Rik
- Chromosome Location
- Chr. 7: 7212996 - 7278289 on Build GRCm38
- UniGene ID
- Mm.387947
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003555.1 |
|
7 |
1493 |
|
GenBank mRNA |
AK018294.1 |
- |
7 |
1431 |
|
AK039785.1 |
- |
1 |
3356 |
|
View Full Product Details