Gene Transcripts

Gene Symbol
Nkx6-2
Entrez Gene ID
14912
Gene Name
NK6 homeobox 2
Gene Aliases
Gtx, Nkx6.2
Chromosome Location
Chr. 7: 139579376 - 139582797 on Build GRCm38
UniGene ID
Mm.28308
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027857.1 4 1740
GenBank mRNA AK008173.1 - 4 1742
BF457051.1 - 1 73
BY245395.1 - 2 244

View Full Product Details