Gene Transcripts
- Gene Symbol
- Malat1
- Entrez Gene ID
- 72289
- Gene Name
- metastasis associated lung adenocarcinoma transcript 1 (non-coding RNA)
- Gene Aliases
- 2210401K01Rik, 9430072K23Rik, AI647968, Neat2
- Chromosome Location
- Chr. 19: 5795690 - 5802671 on Build GRCm38
- UniGene ID
- Mm.298256
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002847.2 |
|
1 |
4684 |
|
GenBank mRNA |
EF177380.1 |
- |
1 |
4684 |
|
FJ209304.1 |
- |
1 |
4684 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AY722410.1 |
- |
1 |
4684 |
|
Chr. - - [-] |
GenBankmRNA |
AI786926.1 |
- |
1 |
309 |
|
Chr. 19 - 5797757 - 5798296 [-] |
View Full Product Details