Gene Transcripts

Gene Symbol
Ppp2r3d
Entrez Gene ID
19054
Gene Name
protein phosphatase 2 (formerly 2A), regulatory subunit B'', delta
Gene Aliases
PP2A B''-PR59, PP2A PR59, PR59, Ppp2r3, Ppp2r3a
Chromosome Location
Chr. 9: 124438769 - 124441010 on Build GRCm38
UniGene ID
Mm.271249
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028110.2 1021
NR_102716.1 1139
GenBank mRNA AK054122.1 - 1021
AK143212.1 - 925
BC096544.1 - 328 6834351

View Full Product Details