Gene Transcripts

Gene Symbol
2010204K13Rik
Entrez Gene ID
68355
Gene Name
RIKEN cDNA 2010204K13 gene
Gene Aliases
0610010M13Rik, Cip7
Chromosome Location
Chr. X: 7411817 - 7422988 on Build GRCm38
UniGene ID
Mm.45111
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027924.1 2 472
NR_027925.1 2 482
GenBank mRNA AF038507.1 - 2 436
AK002502.1 - 821
AK008437.1 - 2 473
AK077356.1 - 2 516
BC042200.1 - 2 435
BC061056.1 - 2 483
BC072610.1 - 2 396


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BX514281.1 - 2 485 Chr. X - 7412107 - 7421049 [-]
GenBankmRNA BU604031.1 - 2 486 Chr. X - 7412070 - 7421047 [-]
GenBankmRNA CD768141.1 - 2 523 Chr. X - 7412102 - 7421087 [-]
GenBankmRNA BB618098.1 - 2 516 Chr. X - 7412041 - 7421080 [-]
GenBankmRNA CD768980.1 - 2 489 Chr. X - 7412103 - 7421050 [-]
GenBankmRNA BB027336.2 - 1 316 Chr. X - 7411817 - 7412486 [-]

View Full Product Details