Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_027924.1 | 2 | 135 | ||
NR_027925.1 | 2 | 145 | |||
GenBank mRNA | AF038507.1 | - | 2 | 99 | |
AK002502.1 | - | 484 | |||
AK008437.1 | - | 2 | 136 | ||
AK077356.1 | - | 2 | 179 | ||
BC042200.1 | - | 2 | 98 | ||
BC061056.1 | - | 2 | 146 | ||
BC072610.1 | - | 2 | 59 |