Gene Transcripts
- Gene Symbol
- Mcpt-ps1
- Entrez Gene ID
- 17223
- Gene Name
- mast cell protease, pseudogene 1
- Gene Aliases
- MMCP-(Y)1, Mmcpy1
- Chromosome Location
- Chr. 14: 55949809 - 55952371 on Build GRCm38
- UniGene ID
- Mm.437339
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028284.1 |
|
2, 3 |
158 |
|
GenBank mRNA |
BY328253.1 |
- |
2, 3 |
159 |
|
X78543.1 |
- |
2, 3 |
137 |
|
View Full Product Details