Gene Transcripts
- Gene Symbol
- Vps39
- Entrez Gene ID
- 269338
- Gene Name
- vacuolar protein sorting 39 (yeast)
- Gene Aliases
- A230065P22Rik, AW743070, Vam6, Vam6P, mVam6
- Chromosome Location
- Chr. 2: 120316461 - 120353144 on Build GRCm38
- UniGene ID
- Mm.246131
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027618.1 |
|
2, 3 |
283 |
|
GenBank mRNA |
AK170831.1 |
- |
2, 3 |
273 |
|
BB655861.1 |
- |
2, 3 |
239 |
|
View Full Product Details