Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|---|
RefSeq | NR_015348.1 | 1 | 348 | |||
GenBank mRNA | U20366.1 | - | LIT1565 Hoxa11as |
1 | 348 | |
U20367.1 | - | LIT1564 Hoxa11as |
1 | 509 |