Gene Transcripts
- Gene Symbol
- Rcbtb2
- Entrez Gene ID
- 105670
- Gene Name
- regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 2
- Gene Aliases
- 2610028E02Rik, 2810420M18Rik, AI448345, AW240694, Chc1l, RC/BTB2
- Chromosome Location
- Chr. 14: 73138773 - 73184055 on Build GRCm38
- UniGene ID
- Mm.280068
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033185.1 |
|
1 |
92 |
|
GenBank mRNA |
AK013119.1 |
- |
1 |
169 |
|
BB854398.1 |
- |
1 |
92 |
|
View Full Product Details