Gene Transcripts
- Gene Symbol
- Speer1
- Entrez Gene ID
- 70896
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein 1
- Gene Aliases
- 4921519H07Rik, SPEER-1, Speer1-ps, Speer1-ps1
- Chromosome Location
- Chr. 5: 11340408 - 11346273 on Build GRCm38
- UniGene ID
- Mm.425218
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001586.3 |
|
1 |
60 |
|
GenBank mRNA |
AY151400.1 |
- |
|
61 |
|
- Gene Symbol
- 4933402N22Rik
- Entrez Gene ID
- 545732
- Gene Name
- RIKEN cDNA 4933402N22 gene
- Gene Aliases
- Chromosome Location
- Chr. 5: 11918043 - 11922804 on Build GRCm38
- UniGene ID
- Mm.389066
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK077088.1 |
- |
|
129 |
|
View Full Product Details