Gene Transcripts
- Gene Symbol
- Rny3
- Entrez Gene ID
- 19874
- Gene Name
- RNA, Y3 small cytoplasmic (associated with Ro protein)
- Gene Aliases
- Chromosome Location
- Chr. 6: 47781624 - 47781724 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024202.2 |
|
1 |
44 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
U84673.1 |
- |
1 |
29 |
|
Chr. - - [-] |
View Full Product Details