Gene Transcripts
- Gene Symbol
- Daw1
- Entrez Gene ID
- 71227
- Gene Name
- dynein assembly factor with WDR repeat domains 1
- Gene Aliases
- 4930563E19Rik, 4933429D11Rik, Wdr69, b2b1116Clo
- Chromosome Location
- Chr. 1: 83159742 - 83210577 on Build GRCm38
- UniGene ID
- Mm.256482
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027636.1 |
|
7 |
651 |
|
GenBank mRNA |
AK016201.1 |
- |
7 |
646 |
|
BU612798.1 |
- |
7 |
614 |
|
View Full Product Details