Gene Transcripts
- Gene Symbol
- Mir10a
- Entrez Gene ID
- 723893
- Gene Name
- microRNA 10a
- Gene Aliases
- Mirn10a, miR-10a, mmu-mir-10a
- Chromosome Location
- Chr. 11: 96317165 - 96317274 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029784.1 |
|
1 |
100 |
|
View Full Product Details