Gene Transcripts

Gene Symbol
Pradc1
Entrez Gene ID
73327
Gene Name
protease-associated domain containing 1
Gene Aliases
1700040I03Rik, Pap21
Chromosome Location
Chr. 6: 85446785 - 85451970 on Build GRCm38
UniGene ID
Mm.379221
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028081.1 1 116
GenBank mRNA BG088874.2 - 1 91
CJ075544.1 - 1 116

View Full Product Details